Learn More
Uline Uline Laser Labels - 4 x 2 S-3847R | Fluorescent Red | 1000 EA
Catalog No. NC2116498
Encompass
Heat-resistant adhesive reduces jamming in your laser or inkjet printer, High quality fluorescent laser labels, Easy to format Microsoft Word and PDF Templates, Labels have round corners, Matte finish, Adhesive Permanent acrylic, Can be used with inkjet or laser printers, Not recommended for use on Poly Bags, Meets the requirements of FDA 21 CFR 175.105 for indirect food contact
This item is not returnable. View return policy
Catalog No. NC2116498 Supplier Uline Supplier No. S3847R
Heat-resistant adhesive reduces jamming in your laser or inkjet printer, High quality fluorescent laser labels, Easy to format Microsoft Word and PDF Templates, Labels have round corners, Matte finish, Adhesive Permanent acrylic, Can be used with inkjet or laser printers, Not recommended for use on Poly Bags, Meets the requirements of FDA 21 CFR 175.105 for indirect food contact