Learn More
Sklar Instruments Sklar Black Laser Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Cross-Serrated with Groove, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1.
SDP
Catalog No. 501436566 Shop All Sklar Instruments Products
Encompass_Preferred
Sklar Black Laser Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Cross-Serrated with Groove, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black Mayo-Hegar Needle Holders are widely used in general surgery for holding medium to large, heavy suture needles. The heavy jaws have cross-serrations and a groove. This instrument is available in several lengths. The shorter length instrument may be used for procedures such as suturing the uterine cervix, while longer lengths may be used for deep tissue suturing. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
This item is not returnable. View return policy
Catalog No. 50-143-6566 Supplier Sklar Instruments Supplier No. 902287
Sklar Black Laser Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Cross-Serrated with Groove, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black Mayo-Hegar Needle Holders are widely used in general surgery for holding medium to large, heavy suture needles. The heavy jaws have cross-serrations and a groove. This instrument is available in several lengths. The shorter length instrument may be used for procedures such as suturing the uterine cervix, while longer lengths may be used for deep tissue suturing. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.