Learn More
GSC Laser Optics Kit
SCI_ED
Catalog No. S39014 Shop All GSC Products
Click to view available options
Grade:
4
Catalog No. Grade
S39014 4
1 options
Catalog No. S39014 Supplier GSC Supplier No. LSOPT7
GSC Laser Optics Kit is ideal for studying optics and the ways light is reflected, absorbed, or transmitted when it encounters various obstacles
  • The included laser ray box can shoot either one, three, or five parallel beams (650nm) across the provided protractor into a number of obstacle accessories to measure how the light reacts and changes paths
  • Included accesories:
    • 1 Circular shape
    • 1 Semi-circle shape
    • 1 Rectangular shape
    • 1 Acrylic bar
    • 1 Equilateral triangle
    • 1 Right triangle
    • 1  trapezoid
    • 1 Bi-concave shape
    • 1 Plano-convex shape
    • 2 Plano-concave shapes
    • 6 Bi-convex shapes
    • 1 Acrylic mirror
    • 1 Equilateral prism (50mm)
    • 1 Acrylic tank (125mm)
Specifications
Grade 4
Age 12
Includes Storage Case, Laser Ray Box, Refraction Tank, 6 lenses
Quantity 1
Dimensions (L x W x H) 19 x 23 x 5 in.
Product Type Education Light
Description Laser Optics Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov