Learn More
ARBOR SCIENTIFIC LASER POINTER VIOLET
SDP
Catalog No. NC0244357
Encompass
Violet, the newest laser pointer color, has a stunning blue-purple glow at 401nm and is used in spectroscopy, fluorescence, microscopy, and more! This violet laser pointer includes two AAA batteries, and a one year warranty.
Catalog No. NC0244357 Supplier ARBOR SCIENTIFIC Supplier No. P27678
Violet, the newest laser pointer color, has a stunning blue-purple glow at 401nm and is used in spectroscopy, fluorescence, microscopy, and more! This violet laser pointer includes two AAA batteries, and a one year warranty.