Learn More
Med Vet International Jorvet Staight Handpiece, for CO2 Laser System, J0941M
Catalog No. 5000495872
Encompass_Preferred
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.
Catalog No. 50-004-95872 Supplier Med Vet International Supplier No. JOR941M
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.