Learn More
Med Vet International Jorvet Ceramic Laser Tip, 10mm x 0.2mm, J0941C, Each
Catalog No. 5000495139
Encompass_Preferred
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.
Catalog No. 50-004-95139 Supplier Med Vet International Supplier No. JOR941C
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.