Learn More
Sigma Aldrich Fine Chemicals Biosciences Laser Tough-SpotsR diam. 3
Catalog No. 502901877
Encompass
Tough-Spots are chemically inert polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths conventional and cryogenic freezer temperatures most organic solvents caustic agents and other challenges without peeling becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Catalog No. 50-290-1877 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T13161PAK
Tough-Spots are chemically inert polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths conventional and cryogenic freezer temperatures most organic solvents caustic agents and other challenges without peeling becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.