Learn More
Cayman Chemical LasofoxIfen 10mg
SDP
Catalog No. 502985971
Encompass_Preferred
A selective estrogen receptor modulator, selectively binds to human ERα (IC50 = 1.5 nM); inhibits bone loss in ovariectomized rats; in clinical studies 0.5 mg/day associated with reduced risks of nonvertebral and vertebral fractures, ER-positive breast cancer, coronary heart disease, and stroke and increased risk of venous thromboembolic events; an inverse agonist at the CB2 cannabinoid receptor
Catalog No. 50-298-5971 Supplier Cayman Chemical Supplier No. 2175410 MG
A selective estrogen receptor modulator, selectively binds to human ERα (IC50 = 1.5 nM); inhibits bone loss in ovariectomized rats; in clinical studies 0.5 mg/day associated with reduced risks of nonvertebral and vertebral fractures, ER-positive breast cancer, coronary heart disease, and stroke and increased risk of venous thromboembolic events; an inverse agonist at the CB2 cannabinoid receptor