Learn More
Apexbio Technology LLC Lasofoxifene (tartrate) 190791-29-8 10mg
SDP
Catalog No. 5000091249
Encompass_Preferred
Lasofoxifene tartrate (CAS 190791-29-8) is a small-molecule selective estrogen receptor modulator (SERM) targeting estrogen receptor-alpha (ER- ) and acting as a selective inverse agonist at the cannabinoid receptor subtype CB2 It is designed to modulate estrogen receptor signaling in a tissue-dependent manner thereby regulating estrogen-related biological pathways Lasofoxifene tartrate exerts its biological activity through dual estrogen agonist antagonist actions depending on tissue context In preclinical studies using ovariectomized rat models Lasofoxifene tartrate prevents estrogen deficiency-associated bone loss reduces serum cholesterol and body mass without inducing uterine hypertrophy Based on these pharmacological properties Lasofoxifene tartrate holds research potential in the management of osteoporosis breast cancer and cardiovascular health in postmenopausal women
Catalog No. 50-000-91249 Supplier Apexbio Technology LLC Supplier No. C563710
Lasofoxifene tartrate (CAS 190791-29-8) is a small-molecule selective estrogen receptor modulator (SERM) targeting estrogen receptor-alpha (ER- ) and acting as a selective inverse agonist at the cannabinoid receptor subtype CB2 It is designed to modulate estrogen receptor signaling in a tissue-dependent manner thereby regulating estrogen-related biological pathways Lasofoxifene tartrate exerts its biological activity through dual estrogen agonist antagonist actions depending on tissue context In preclinical studies using ovariectomized rat models Lasofoxifene tartrate prevents estrogen deficiency-associated bone loss reduces serum cholesterol and body mass without inducing uterine hypertrophy Based on these pharmacological properties Lasofoxifene tartrate holds research potential in the management of osteoporosis breast cancer and cardiovascular health in postmenopausal women