Learn More
Cayman Chemical Lasofoxifene (tartrate), Purity: ≥98%, CAS: 190791-29-8, 50 mg
SDP
Catalog No. 502966562
Encompass_Preferred
A third-generation SERM that selectively binds to human ERα (IC50 = 1.5 nM) and inhibits bone loss in ovariectomized rats; used in the treatment of postmenopausal osteoporosis and ER-positive breast cancer; also acts as an inverse agonist at the CB2 cannabinoid receptor
Catalog No. 50-296-6562 Supplier Cayman Chemical Supplier No. 1764650 MG
A third-generation SERM that selectively binds to human ERα (IC50 = 1.5 nM) and inhibits bone loss in ovariectomized rats; used in the treatment of postmenopausal osteoporosis and ER-positive breast cancer; also acts as an inverse agonist at the CB2 cannabinoid receptor