Learn More
Electron Microscopy Sciences Tough-Spots Labels on Laser Sheets, 3/8", Blue
SDP
Catalog No. 501900046
Encompass_Preferred
Pre-cut, peel-off labels for microcentrifuge tube top on sheets. Withstands freezer temperatures down to 196 degree C (liquid and vapor phase nitrogen), boiling water baths (100 degree C), autoclaving and moderate oven temperatures (150 degree C). Chemically inert labels resist most organic solvents and caustic agents. They adhere to plastics, glass and metals without cracking, peeling or degrading.
Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam laser printers.
Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations, resulting in a strong adhesive bond.
Size: 3/8"
Color: Blue
Catalog No. 50-190-0046 Supplier Electron Microscopy Sciences Supplier No. 77585B
Pre-cut, peel-off labels for microcentrifuge tube top on sheets. Withstands freezer temperatures down to 196 degree C (liquid and vapor phase nitrogen), boiling water baths (100 degree C), autoclaving and moderate oven temperatures (150 degree C). Chemically inert labels resist most organic solvents and caustic agents. They adhere to plastics, glass and metals without cracking, peeling or degrading.
Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam laser printers.
Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations, resulting in a strong adhesive bond.
Size: 3/8"
Color: Blue