Learn More
Aviva Systems Biology Corp LASS2 Antibody - N-terminal region (ARP31922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LASS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LASS2. Alternate names: CerS2, FLJ10243, L3, MGC987, SP260, TMSG1, LASS2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54373