Learn More
Aviva Systems Biology Corp LASS5 Antibody - N-terminal region (ARP37810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LASS5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LASS5. Alternate names: CerS5, FLJ25304, MGC45411, Trh4, LASS5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-56438