Learn More
Medchemexpress LLC LASSBio-1135 | 852453-71-5 | 99.1% | 286.33 g·mol^-1 | C18H14N4 | 5 MG
SDP
Catalog No. 5000262829
Encompass_Preferred
LASSBio-1135 is an orally active TRPV1 antagonist with anti-inflammatory and analgesic activity. It moderately inhibits human PGHS-2 (IC50 = 18.5 μM) and has shown in vivo efficacy by reversing capsaicin-induced thermal hyperalgesia and reducing carrageenan-induced paw edema.
  • Orally active TRPV1 antagonist.
  • Moderate inhibition of human PGHS-2 (IC50 = 18.5 μM).
  • Demonstrated in vivo efficacy in pain and inflammation models.
  • High purity suitable for research applications.
  • Molecular weight 286.33 g·mol^-1 and formula C18H14N4.
  • Available in small research quantities for dosing studies.
  • Stable when stored as powder at -20°C for extended periods.
Catalog No. 50-002-62829 Supplier Medchemexpress LLC Supplier No. HY1319865MG
LASSBio-1135 is an orally active TRPV1 antagonist with anti-inflammatory and analgesic activity. It moderately inhibits human PGHS-2 (IC50 = 18.5 μM) and has shown in vivo efficacy by reversing capsaicin-induced thermal hyperalgesia and reducing carrageenan-induced paw edema.
  • Orally active TRPV1 antagonist.
  • Moderate inhibition of human PGHS-2 (IC50 = 18.5 μM).
  • Demonstrated in vivo efficacy in pain and inflammation models.
  • High purity suitable for research applications.
  • Molecular weight 286.33 g·mol^-1 and formula C18H14N4.
  • Available in small research quantities for dosing studies.
  • Stable when stored as powder at -20°C for extended periods.