Learn More
Medchemexpress LLC Lassbio-1135 | 852453-71-5 | 99.1% | C18H14N4 | 10MG
SDP
Catalog No. 5000261119
Encompass_Preferred
LASSBio-1135 is an orally active TRPV1 antagonist and anti-inflammatory/analgesic research compound. It shows moderate inhibition of human PGHS-2 (IC50 = 18.5 μM) and has demonstrated in vivo efficacy reversing capsaicin-induced thermal hyperalgesia and reducing carrageenan-induced paw edema. The material is supplied as a white to off-white solid with reported high purity and recommended low-temperature storage for stability.
  • orally active TRPV1 antagonist with anti-inflammatory and analgesic activity
  • moderate inhibition of human PGHS-2 (IC50 = 18.5 μM)
  • demonstrated in vivo efficacy in pain and inflammation models
  • high purity solid, white to off-white appearance
  • recommended storage at low temperatures for long-term stability
Catalog No. 50-002-61119 Supplier Medchemexpress LLC Supplier No. HY13198610MG
LASSBio-1135 is an orally active TRPV1 antagonist and anti-inflammatory/analgesic research compound. It shows moderate inhibition of human PGHS-2 (IC50 = 18.5 μM) and has demonstrated in vivo efficacy reversing capsaicin-induced thermal hyperalgesia and reducing carrageenan-induced paw edema. The material is supplied as a white to off-white solid with reported high purity and recommended low-temperature storage for stability.
  • orally active TRPV1 antagonist with anti-inflammatory and analgesic activity
  • moderate inhibition of human PGHS-2 (IC50 = 18.5 μM)
  • demonstrated in vivo efficacy in pain and inflammation models
  • high purity solid, white to off-white appearance
  • recommended storage at low temperatures for long-term stability