Learn More
Microsolv Technology Corporation Mobile Phase Filter, 10um, Last Drop™, PTFE, 1/8" Connector. 1 EA.
Catalog No. 502271593
Encompass_Preferred
These Last Drop, PEEK and Polypropylene Mobile Phase Filters are manufactured with a 10um PTFE Filter Element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to "vacuum" up to the "last drop of liquid" in the reservoir bottle. The all PEEK, 1/8" Tubing Connector uses the fitting and ETFE ferrule to hold non-flanged low pressure tubing in the Filter without leaking. The 10um pore size Element will filter undissolved particles from your solvents that are 10um or larger preventing them from damaging your HPLC pumps, columns or instrument. Install these filters directly in your mobile phase bottles.
This item is not returnable. View return policy
Catalog No. 50-227-1593 Supplier Microsolv Technology Corporation Supplier No. 5884609
These Last Drop, PEEK and Polypropylene Mobile Phase Filters are manufactured with a 10um PTFE Filter Element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to "vacuum" up to the "last drop of liquid" in the reservoir bottle. The all PEEK, 1/8" Tubing Connector uses the fitting and ETFE ferrule to hold non-flanged low pressure tubing in the Filter without leaking. The 10um pore size Element will filter undissolved particles from your solvents that are 10um or larger preventing them from damaging your HPLC pumps, columns or instrument. Install these filters directly in your mobile phase bottles.