Learn More
Microsolv Technology Corporation Mobile Phase Filter, 5.0um, Last Drop™, PTFE, 1/16" Connector. 1 EA.
Supplier:  Microsolv Technology Corporation 5884604
Encompass_Preferred
These Last Drop, PEEK and Polypropylene Mobile Phase Filters are manufactured with a 5um PTFE Filter Element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to "vacuum" up to the "last drop of liquid" in the reservoir bottle. The all PEEK, 1/16" Tubing Connector uses the fitting and an ETFE ferrule to hold non-flanged low pressure tubing in to the Filter without leaking. The 5um pore size Element will filter undissolved particles from your solvents that are 5um or larger preventing them from damaging your HPLC pumps, columns or instrument. Install these filters directly in your mobile phase bottles.
Catalog No. 50-227-1589