Learn More
Microsolv Technology Corporation Last Drop Fltr 2.5Um Ptfe 1/8
Supplier:  Microsolv Technology Corporation 5884603
Encompass_Preferred
Mobile Phase Filter, 2.5um, Last Drop, PTFE, 1/8"" Connector. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52655