Learn More
Microsolv Technology Corporation Last Drop Fltr 2Um Ss W/Adapt
Catalog No. 5001452652
Encompass_Preferred
Mobile Phase Filter, 2um, Last Drop, Stainless Steel, Tripod Tubing Connector. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52652 Supplier Microsolv Technology Corporation Supplier No. 5883501
Mobile Phase Filter, 2um, Last Drop, Stainless Steel, Tripod Tubing Connector. MicroSolv Brand. 1/EA.