Learn More
Medchemexpress LLC HY-108540 50mg Medchemexpress, LAT1-IN-1 CAS:20448-79-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10854050MG
Encompass_Preferred
Medchemexpress, HY-108540 50mg LAT1-IN-1 CAS:20448-79-7 LAT1-IN-1 (BCH) is a selective and competitive inhibitor of large neutral amino acid transporter 1 (LAT1) significantly inhibit cellular uptake of amino acids and mTOR phosphorylation, which induces the suppression of cancer growth and apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0746