Learn More
Medchemexpress LLC Latanoprost | 130209-82-4 | 216074 | >99.85% | 100 MG
SDP
Catalog No. 5000408271
Encompass_Preferred
Latanoprost is a prostaglandin F2α analogue primarily utilized in glaucoma research. This compound is capable of penetrating the cornea, where it is subsequently hydrolyzed by esterase into its active form, latanoprost acid. The acid then functions as an F-prostaglandin (FP) receptor agonist, efficiently reducing intraocular pressure by facilitating an increased outflow of aqueous humor via the uvea.
  • Prostaglandin F2α analogue for research
  • Passes through the cornea and is hydrolyzed to active acid form
  • Acts as an F-prostaglandin (FP) receptor agonist
  • Effectively lowers intraocular pressure (IOP)
  • Increases aqueous humor outflow through the uvea
Catalog No. 50-004-08271 Supplier Medchemexpress LLC Supplier No. HYB0577100MG
Latanoprost is a prostaglandin F2α analogue primarily utilized in glaucoma research. This compound is capable of penetrating the cornea, where it is subsequently hydrolyzed by esterase into its active form, latanoprost acid. The acid then functions as an F-prostaglandin (FP) receptor agonist, efficiently reducing intraocular pressure by facilitating an increased outflow of aqueous humor via the uvea.
  • Prostaglandin F2α analogue for research
  • Passes through the cornea and is hydrolyzed to active acid form
  • Acts as an F-prostaglandin (FP) receptor agonist
  • Effectively lowers intraocular pressure (IOP)
  • Increases aqueous humor outflow through the uvea