Learn More
Apexbio Technology LLC Latanoprost 130209-82-4 50mg
SDP
Catalog No. 5000063779
Encompass_Preferred
Latanoprost (CAS 130209-82-4) is a synthetic analogue of prostaglandin F2 that functions primarily as an agonist at the prostaglandin FP receptor Activation of this receptor enhances aqueous humor drainage through the uveoscleral outflow pathway in ocular tissues resulting in reduced intraocular pressure In research settings latanoprost is employed to study prostaglandin-mediated signaling pathways and serves as a standard reference agent in glaucoma and ocular hypertension models
Catalog No. 50-000-63779 Supplier Apexbio Technology LLC Supplier No. B341150
Latanoprost (CAS 130209-82-4) is a synthetic analogue of prostaglandin F2 that functions primarily as an agonist at the prostaglandin FP receptor Activation of this receptor enhances aqueous humor drainage through the uveoscleral outflow pathway in ocular tissues resulting in reduced intraocular pressure In research settings latanoprost is employed to study prostaglandin-mediated signaling pathways and serves as a standard reference agent in glaucoma and ocular hypertension models