Learn More
Selleck Chemical LLC Latanoprost 5mg 130209-82-4 Xalatan, PhXA41, PHXA-41
SDP
Supplier:  Selleck Chemical LLC S47095MG
Encompass_Preferred
Latanoprost (Xalatan, PhXA41) is a prostaglandin F2alpha analogue and a prostanoid selective FP receptor agonist with an ocular hypertensive effect. Latanoprost increases uveoscleral outflow and thereby reduces intraocular pressure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4212