Learn More
Medchemexpress LLC Latanoprost-d4 | 130209-82-4 | 98.0% | 500 UG
SDP
Catalog No. 5000408673
Encompass_Preferred
Latanoprost-d4 is the deuterium-labeled Latanoprost, a prostaglandin F2α analogue and an agonist for the FP prostanoid receptor, which lowers intraocular pressure (IOP). It serves as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs, offering valuable insights in research.
  • Deuterium labeled compound
  • Prostaglandin F2α analogue
  • Agonist for FP prostanoid receptor
  • Lowers intraocular pressure (IOP)
  • Useful as a tracer
  • Functions as an internal standard for quantitative analysis
  • Can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08673 Supplier Medchemexpress LLC Supplier No. HYB0577S500UG
Latanoprost-d4 is the deuterium-labeled Latanoprost, a prostaglandin F2α analogue and an agonist for the FP prostanoid receptor, which lowers intraocular pressure (IOP). It serves as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs, offering valuable insights in research.
  • Deuterium labeled compound
  • Prostaglandin F2α analogue
  • Agonist for FP prostanoid receptor
  • Lowers intraocular pressure (IOP)
  • Useful as a tracer
  • Functions as an internal standard for quantitative analysis
  • Can affect pharmacokinetic and metabolic profiles of drugs