Learn More
Cayman Chemical Latanoprostd4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10006556500 UG
Encompass_Preferred
An internal standard for the quantification of latanoprost by GC- or LC-MS
Catalog No. 50-288-7767