Learn More
Cayman Chemical Latanoprostd4 GC MS Standards
SDP
Catalog No. 502891098
Encompass_Preferred
An internal standard for the quantification of latanoprost by GC- or LC-MS
Catalog No. 50-289-1098 Supplier Cayman Chemical Supplier No. 10006556100 UG
An internal standard for the quantification of latanoprost by GC- or LC-MS