Learn More
Cayman Chemical Latanoprost-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502887761
Encompass_Preferred
An internal standard for the quantification of latanoprost by GC- or LC-MS
Catalog No. 50-288-7761 Supplier Cayman Chemical Supplier No. 100065561 MG
An internal standard for the quantification of latanoprost by GC- or LC-MS