Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Latanoprostene bunod | 860005-21-6 | MFCD30532608 | ≥95.0% | 507.62 g/mol | C27H41NO8 | 10 MG
SDP

Catalog No. 5000298358
Encompass_Preferred

Latanoprostene bunod is a nitric oxide-donating prostaglandin F2α analog developed as a topical ophthalmic agent to reduce intraocular pressure. Supplied for research and analytical use, it is provided as a pure solid with recommended cold storage and is commonly used in pharmacology, formulation, and analytical method development.

  • Nitric oxide-donating prostaglandin F2α analog.
  • Reduces intraocular pressure in ocular hypertension models.
  • Supplied as a solid analytical standard for research use.
  • Available in 1 mg, 5 mg, and 10 mg pack sizes.
  • CAS number 860005-21-6; formula C27H41NO8; MW 507.62 g/mol.
  • Store at -20 °C for long-term stability; short-term storage at 4 °C is acceptable.

Catalog No. 50-002-98358 Supplier Medchemexpress LLC Supplier No. HY1951810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Latanoprostene bunod is a nitric oxide-donating prostaglandin F2α analog developed as a topical ophthalmic agent to reduce intraocular pressure. Supplied for research and analytical use, it is provided as a pure solid with recommended cold storage and is commonly used in pharmacology, formulation, and analytical method development.

  • Nitric oxide-donating prostaglandin F2α analog.
  • Reduces intraocular pressure in ocular hypertension models.
  • Supplied as a solid analytical standard for research use.
  • Available in 1 mg, 5 mg, and 10 mg pack sizes.
  • CAS number 860005-21-6; formula C27H41NO8; MW 507.62 g/mol.
  • Store at -20 °C for long-term stability; short-term storage at 4 °C is acceptable.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.