Description
- Offers enhanced performance compared to traditional luminol, is easy to use, and does not interfere with DNA analysis.
- Convenient tablet form enables quick preparation and easy application in the field.
- Mix in a spray bottle containing 125 mL of distilled water
- Does not adversely affect DNA
- Makes invisible bloodstains produce an intense blue chemiluminescence odorless
- Fully soluble
Specifications
Specifications
| Product Type | Bloodstain Reagent |
| For Use With (Application) | Latent Bloodstain Detection |
| Quantity | 4/Pk. |