Learn More
Mopec Latent Print Powder Magnetic Wand
SDP
Catalog No. 22446247 Shop All Mopec Products
Change view
Click to view available options
Includes:
1 Disposable Cover, 1 stem
3 Disposable Covers, 1 Stem
5 Disposable Covers, 1 Stem
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-446-247 5 Disposable Covers, 1 Stem
22-446-219 1 Disposable Cover, 1 stem
22-446-205 3 Disposable Covers, 1 Stem
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-247 Supplier Mopec Supplier No. FS023
Add to Cart
Add to Cart
The TEX system is a simple, reliable solution for collecting latent prints while helping prevent Cross-contamination between scenes.
  • Lightweight magnetic wand
  • 1 Stem, various amounts of covers
  • Reduces Cross-contamination between collection sites
  • Enables consistent latent print development
  • Options and accessories include the FS024 10-cover pack and the FS025 25-cover pack.
  • Its disposable cover design ensures cleaner evidence handling, making it a practical tool for maintaining DNA sample integrity during investigations
Specifications
Product Type Magnetic Wand
Includes 5 Disposable Covers, 1 Stem