Learn More
Medchemexpress LLC Latent TGF beta 1/TG 100ug
SDP
Catalog No. 5000179970
Encompass_Preferred
Latent TGF beta 1 (latent TGFB1) is the inactive form of TGF-B1 Latent TGF beta 1 associates with the extracellular matrix (ECM) via LTBP LTBPs are components of the ECM so that the proteolytic cleavage of LTBP can lead to the release of latent TGF-beta 1 from the matrix Besides the proteolytic cleavage and liberation of active TGFB1 is performed by BMP-1 by a variety of matrix metalloproteinases (MMPs)[1][3] Latent TGF beta 1/TGFB1 Protein Rat (HEK293 His) is the recombinant rat-derived Latent TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79970 Supplier Medchemexpress LLC Supplier No. HYP73615100UG
Latent TGF beta 1 (latent TGFB1) is the inactive form of TGF-B1 Latent TGF beta 1 associates with the extracellular matrix (ECM) via LTBP LTBPs are components of the ECM so that the proteolytic cleavage of LTBP can lead to the release of latent TGF-beta 1 from the matrix Besides the proteolytic cleavage and liberation of active TGFB1 is performed by BMP-1 by a variety of matrix metalloproteinases (MMPs)[1][3] Latent TGF beta 1/TGFB1 Protein Rat (HEK293 His) is the recombinant rat-derived Latent TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag