Learn More
Medchemexpress LLC Latent TGF beta 1/TG 10ug | 10ug
SDP
Catalog No. 5000187388
Encompass_Preferred
Transforming growth factor beta-1 proprotein is the precursor of the Latency-associated peptide (LAP) and Transforming growth factor beta-1 (TGF-beta-1) chains TGF beta 1/TGFB1 Protein Mouse (HEK293 His) is a recombinant protein (L30-S390) produced by HEK293 cells with His tag[1][2]
Catalog No. 50-001-87388 Supplier Medchemexpress LLC Supplier No. HYP7836010UG
Transforming growth factor beta-1 proprotein is the precursor of the Latency-associated peptide (LAP) and Transforming growth factor beta-1 (TGF-beta-1) chains TGF beta 1/TGFB1 Protein Mouse (HEK293 His) is a recombinant protein (L30-S390) produced by HEK293 cells with His tag[1][2]