Learn More
Medchemexpress LLC Latent TGF beta 1/TG 50ug
SDP
Catalog No. 5000180463
Encompass_Preferred
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha Latent TGF beta 1/TGFB1 Protein Human (HEK293 His) is the recombinant human-derived Latent TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80463 Supplier Medchemexpress LLC Supplier No. HYP7342650UG
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha Latent TGF beta 1/TGFB1 Protein Human (HEK293 His) is the recombinant human-derived Latent TGF beta 1/TGFB1 protein expressed by HEK293 with C-His labeled tag