Learn More
Medchemexpress LLC Latent TGF beta 1/TG 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7858450UG
Encompass_Preferred
Latent TGF beta 1/TGFB1 Protein is a large extracellular matrix protein and an associated ligand of fibrillinmicrofibrils Latent TGF beta 1/TGFB1 Protein Cynomolgus (C33S HEK293 His) is a recombinant protein (L30-S390 C33S) produced by HEK293 cells with His tag[1][2]
Catalog No. 50-001-94306