Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Latent transforming growth factor beta-3 (human) | >95.0% | 100UG
SDP

Catalog No. 5000174942
Encompass_Preferred

Recombinant human latent transforming growth factor beta-3 (latent TGF-β3) expressed in HEK293 cells and supplied as a lyophilized protein containing the latency-associated peptide. The product is formulated from filtered PBS with trehalose as a stabilizer and is intended for research applications involving TGF-β3 signaling, activation studies, and extracellular matrix interactions.

  • Expressed in HEK293 cells with an N-terminal His tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular weight 45.9 kDa; observed ~46-60 kDa by SDS-PAGE.
  • Lyophilized formulation from 0.22 μm filtered PBS with trehalose protectant.
  • Stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Catalog No. 50-001-74942 Supplier Medchemexpress LLC Supplier No. HYP700890100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human latent transforming growth factor beta-3 (latent TGF-β3) expressed in HEK293 cells and supplied as a lyophilized protein containing the latency-associated peptide. The product is formulated from filtered PBS with trehalose as a stabilizer and is intended for research applications involving TGF-β3 signaling, activation studies, and extracellular matrix interactions.

  • Expressed in HEK293 cells with an N-terminal His tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular weight 45.9 kDa; observed ~46-60 kDa by SDS-PAGE.
  • Lyophilized formulation from 0.22 μm filtered PBS with trehalose protectant.
  • Stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.