Learn More
ACROBiosystems ACROBiosystems Human Latent TGF-beta 1 / TGFB1 Protein, His Tag
Catalog No. 501957305
Encompass_Preferred
Recombinant Protein;1MG;Human Latent TGFB1, His Tag (TG1-H524x) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P01137-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42.6 kDa (monomer). The protein migrates as 37-45 kDa (LAP) and 13 kDa (TGFB1) under reducing (R) condition (SDS-PAGE) due to glycosylation and Interchain disulfide bond.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Human LRRC32, Fc Tag (Cat. No. LR2-H5256) with a linear range of 0.039-0.625 μg/mL (Routinely tested).;Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.2
Catalog No. 50-195-7305 Supplier ACROBiosystems Supplier No. TG1H524X1MG
Recombinant Protein;1MG;Human Latent TGFB1, His Tag (TG1-H524x) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P01137-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42.6 kDa (monomer). The protein migrates as 37-45 kDa (LAP) and 13 kDa (TGFB1) under reducing (R) condition (SDS-PAGE) due to glycosylation and Interchain disulfide bond.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Human LRRC32, Fc Tag (Cat. No. LR2-H5256) with a linear range of 0.039-0.625 μg/mL (Routinely tested).;Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.2