Learn More
Biochain Institute Inc Rapid Gold Immuno-assay Kit for Antibody IgG to Tuberculosis (TB), 1 kit/PK
SDP
Catalog No. 50181642
Encompass_Preferred
BioChain?€?s Rapid Gold Immuno-assay Kit for tuberculosis is a fast and easy test which uses purified recombinant TB antigen and gold-labeled conjugate coated on the surface of a nitrocellulose membrane. Plasma or serum samples are added and the interaction of the antigen-antibody complex and gold-labeled conjugate will produce a pink color on the membrane. The intensity of this color will reflect the antibody intensity in the sample.
Catalog No. 50-181-642 Supplier Biochain Institute Inc Supplier No. Z7010010
BioChain?€?s Rapid Gold Immuno-assay Kit for tuberculosis is a fast and easy test which uses purified recombinant TB antigen and gold-labeled conjugate coated on the surface of a nitrocellulose membrane. Plasma or serum samples are added and the interaction of the antigen-antibody complex and gold-labeled conjugate will produce a pink color on the membrane. The intensity of this color will reflect the antibody intensity in the sample.