Learn More
MOLECULAR DEPOT LLC Latex Activation - 100 mL
Catalog No. 5000013362
Encompass_Preferred
Microparticle Activation Buffer (100 mL) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to dilute particles from stock solutions and activate particles for covalent linkage (such as NHS and EDC activation) and wash particles after activation and before conjugation For in vitro research use only
Catalog No. 50-000-13362 Supplier MOLECULAR DEPOT LLC Supplier No. B2010003S
Microparticle Activation Buffer (100 mL) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to dilute particles from stock solutions and activate particles for covalent linkage (such as NHS and EDC activation) and wash particles after activation and before conjugation For in vitro research use only