Learn More
MOLECULAR DEPOT LLC Latex Activation Buffer - 1 L
Catalog No. 5000013361
Encompass_Preferred
Microparticle Activation Buffer (1 L) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to dilute particles from stock solutions activate particles for covalent linkage (such as NHS and EDC activation) and wash particles after activation and before conjugation For in vitro research use only
Catalog No. 50-000-13361 Supplier MOLECULAR DEPOT LLC Supplier No. B2010003L
Microparticle Activation Buffer (1 L) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to dilute particles from stock solutions activate particles for covalent linkage (such as NHS and EDC activation) and wash particles after activation and before conjugation For in vitro research use only