Learn More
Electron Microscopy Sciences Latex Finger Cots Latex Finger Cots Medium
SDP
Catalog No. 50930838
Encompass_Preferred
Inexpensive, expandable finger cots. Seamless. Protective dexterity without covering the entire hand. Powderless, non-rolled. 3 1/2 mil (0.0030") and 2 3/4"(40 mm) long.
This item is not returnable. View return policy
Catalog No. 50-930-838 Supplier Electron Microscopy Sciences Supplier No. 71131
Inexpensive, expandable finger cots. Seamless. Protective dexterity without covering the entire hand. Powderless, non-rolled. 3 1/2 mil (0.0030") and 2 3/4"(40 mm) long.