Learn More
Description
- No certificate of analysis is available
- Health Risk: 1
- Flammability: 0
- Reactivity: 1
Specifications
Specifications
| Chemical Name or Material | Ammonium hydroxide |
| Color | Red |
| Physical Form | Liquid |
| CAS | 1336-21-6 |
| Molecular Formula | H5NO |
| MDL Number | MFCD00066650 |
| Quantity | 4 L |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| SMILES | N.O |
| IUPAC Name | amine hydrate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

