Learn More
Sigma Aldrich Fine Chemicals Biosciences Latex rubber laboratory tu
Catalog No. 502901870
Encompass
High-purity seamless amber tubing for general-purpose use with liquids and gases. Durometer 35 /-5. Talc-free.
Catalog No. 50-290-1870 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2558581PAK
Add to Cart
Add to Cart
High-purity seamless amber tubing for general-purpose use with liquids and gases. Durometer 35 /-5. Talc-free.