Learn More
MOLECULAR DEPOT LLC Latex Stabilizer - 1 L
Supplier:  MOLECULAR DEPOT LLC C1010002L
Encompass_Preferred
Latex Microparticles Stabilizer Solution (1 L) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer is especially effective at stabilizing particles used in immunoturbidimetry Particles are simply spun down (or diafiltered) to exchange their original buffer into this buffer The included proprietary stabilizers can extend particle suspension shelf-life to up to 24 months at 2-8C For in vitro research use only
Catalog No. 50-000-13575