Learn More
Andwin Scientific LATEX TUBING 50 FT.
SDP
Catalog No. NC9518520
Encompass_Preferred
The Amber Latex Rubber Tubing is ideal for all kinds of glass connections where highly elastic, clinging tubing is desired. The tube features a durometer hardness of Shore A, 35 and a tensile strength of 3500 psi. Its wall has a thickness of 3/16" a
This item is not returnable. View return policy
Catalog No. NC9518520 Supplier Andwin Scientific Supplier No. 62996145
The Amber Latex Rubber Tubing is ideal for all kinds of glass connections where highly elastic, clinging tubing is desired. The tube features a durometer hardness of Shore A, 35 and a tensile strength of 3500 psi. Its wall has a thickness of 3/16" a