Learn More
MOLECULAR DEPOT LLC Latex Wash Buffer - 100 mL
Catalog No. 5000013531
Encompass_Preferred
Microparticle Washing Buffer (100 mL) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to wash microparticles following activation with cross-linking groups such as N-Ethyl-N-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDAC) and N-Hydroxysuccinimide (NHS) and conjugation to a target molecule The pH salt and detergent composition of this buffer are essential to counteract non-specific interactions For in vitro research use only
Catalog No. 50-000-13531 Supplier MOLECULAR DEPOT LLC Supplier No. B2010004S
Microparticle Washing Buffer (100 mL) is an optimized versatile buffer for the conjugation of antibodies proteins peptides and small molecules to polystyrene (Latex) microparticles and nanoparticles This buffer can be used to wash microparticles following activation with cross-linking groups such as N-Ethyl-N-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDAC) and N-Hydroxysuccinimide (NHS) and conjugation to a target molecule The pH salt and detergent composition of this buffer are essential to counteract non-specific interactions For in vitro research use only