Learn More
Lath™ Powder Free Nitrile Gloves
SDP
Catalog No. NC1865030
Encompass
Change view
Click to view available options
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
NC1865030 Medium
NC1901843 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NC1865030 Supplier Pantek Technologies LLC Supplier No. 410000237
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Lath™ is the preferred choice for powder free nitrile gloves when thickness and break resistance are a concern.
  • Non-sterile & powder free
  • Made from synthetic nitrile latex
  • Ambidextrous with textured fingers and a beaded cuff
  • Conforms to ASTM D6319 and EN455-1/2/3/4 standards
  • Thickness: 5 mil (palm), 5 mil (fingers)
  • Color: Blue
Specifications
Certifications/Compliance ASTM D6319
Certifications/Compliance (Europe) EN455-1/2/3/4
Color Blue
Disposable Single-use
Finger Thickness (English) 5 mil
Finger Thickness (Metric) 0.13 mm
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Size Medium
Thickness (English) 5 mil
Thickness (Metric) 0.13 mm