Learn More
Grainger Lathe Tool Bit Set 11pc Carbide
Catalog No. 502766009
Encompass_Preferred
The Grainger website features brazed tip lathe tool bits for turning, boring, and grooving operations on lathes. These high-speed steel bits have brazed carbide tips to provide good wear resistance and cutting ability. They come in a variety of shapes including square, round, diamond, and threading profiles. The bits fit standard tool holders and feature positive rake angles for free cutting. They are available in fractional inch sizes ranging from 1/8" to 1/2". Grainger offers a wide selection of brazed tip tool bits from brands like M.A. Ford, Valenite, and Allied Machine & Engineering.
Catalog No. 50-276-6009 Supplier Grainger Supplier No. 16N207
The Grainger website features brazed tip lathe tool bits for turning, boring, and grooving operations on lathes. These high-speed steel bits have brazed carbide tips to provide good wear resistance and cutting ability. They come in a variety of shapes including square, round, diamond, and threading profiles. The bits fit standard tool holders and feature positive rake angles for free cutting. They are available in fractional inch sizes ranging from 1/8" to 1/2". Grainger offers a wide selection of brazed tip tool bits from brands like M.A. Ford, Valenite, and Allied Machine & Engineering.