Learn More
Medchemexpress LLC HY-113486 10mg Medchemexpress, Lathosterol CAS:80-99-9 Purity:>98%
SDP
Catalog No. 501969257
Encompass_Preferred
Medchemexpress, HY-113486 10mg Lathosterol CAS:80-99-9 Lathosterol is a cholesterol-like molecule. Serum Lathosterol concentration is an indicator of whole-body cholesterol synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9257 Supplier Medchemexpress LLC Supplier No. HY11348610MG
Medchemexpress, HY-113486 10mg Lathosterol CAS:80-99-9 Lathosterol is a cholesterol-like molecule. Serum Lathosterol concentration is an indicator of whole-body cholesterol synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.