Learn More
Cayman Chemical Lathosterol, Purity: ≥95%, CAS: 80-99-9, 50 mg
SDP
Catalog No. 502896051
Encompass_Preferred
An intermediate in the biosynthesis of cholesterol; serum levels of lathosterol have been used as biomarkers of excess cholesterol production; accumulates in lathosterolosis
Catalog No. 50-289-6051 Supplier Cayman Chemical Supplier No. 900310250 MG
An intermediate in the biosynthesis of cholesterol; serum levels of lathosterol have been used as biomarkers of excess cholesterol production; accumulates in lathosterolosis