Learn More
Kyfora Bio Lathosterol 5 MG
Catalog No. 502559794
Encompass_Preferred
Lathosterol is a precursor in the cholesterol synthesis pathway. Lathosterol can be used as a biomarker of excess cholesterol production as lathosterol levels in serum corresponds with cholesterol synthesis.
Catalog No. 50-255-9794 Supplier Kyfora Bio Supplier No. 145235
Lathosterol is a precursor in the cholesterol synthesis pathway. Lathosterol can be used as a biomarker of excess cholesterol production as lathosterol levels in serum corresponds with cholesterol synthesis.